Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
6,9-Diamino-2-ethoxyacridine-DL-lactate monohydrate
Synonym(s):
Acrinol, Ethacridine lactate salt, Ethodin
Empirical Formula (Hill Notation):
C
15
H
15
N
3
O · C
3
H
6
O
3
· H
2
O
CAS Number:
6402-23-9
Molecular Weight:
361.39
EC Number:
217-408-1
Products
Product Category
building blocks (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Markush Class
Markush Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
D16606
6,9-Diamino-2-ethoxyacridine-DL-lactate monohydrate,
99%
Expand
View Pricing