5,7-Dihydroxy-4-methylcoumarin
Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing