
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside

Synonym(s):
5,6-Dichloro-1-β-D-ribofuranosylbenzimidazole, 5,6-Dichlorobenzimidazole riboside, DRB
Empirical Formula (Hill Notation):
C12H12Cl2N2O4
CAS Number:
Molecular Weight:
319.14
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing