Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-Methylmorpholine
Synonym(s):
NMM, 4-Methylmorpholine
Empirical Formula (Hill Notation):
C
5
H
11
NO
CAS Number:
109-02-4
Molecular Weight:
101.15
EC Number:
203-640-0
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
M56557
4-Methylmorpholine,
ReagentPlus
®
, 99%
Expand
View Pricing
407704
4-Methylmorpholine,
purified by redistillation, ≥99.5%
Expand
View Pricing
8.05894
4-Methylmorpholine,
for synthesis
Expand
View Pricing
67869
4-Methylmorpholine,
BioXtra, suitable for protein sequencing, ≥99.5% (GC)
Expand
View Pricing
67870
4-Methylmorpholine,
purum, ≥98.0% (GC)
Expand
View Pricing