
4-Bromo-3-thiophenecarboxylic acid

Synonym(s):
4-Bromothiophene-3-carboxylic acid
Empirical Formula (Hill Notation):
C5H3BrO2S
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing