Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3-Nitropropionic acid
Synonym(s):
3-Nitropropionic acid
Linear Formula:
O
2
N(CH
2
)
2
CO
2
H
CAS Number:
504-88-1
Molecular Weight:
119.08
EC Number:
208-003-0
Products
Product Category
bioactive small molecules (2)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
N5636
3-Nitropropionic acid,
≥97% (HPLC), powder or crystals, mitochondrial complex II inhibitor
Expand
View Pricing
164603
3-Nitropropionic acid,
A cell-permeable, irreversible inactivator (~3 mM) of succinate dehydrogenase that upon oxidation by Complex II forms a covalent adduct with Arg297 in the active site of the enzyme.
Expand
View Pricing
N22908
3-Nitropropionic acid,
97%
Expand
View Pricing