Skip to Content
Merck
3-Methoxy-3-methyl-1-butanol ≥98%

3-Methoxy-3-methyl-1-butanol

3-Methoxy-3-methyl-1-butanol ≥98%
Linear Formula:
(CH3)2C(OCH3)CH2CH2OH
CAS Number:
Molecular Weight:
118.17
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing