Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Thiobarbituric acid
Synonym(s):
2-Thiobarbituric acid, 4,6-Dihydroxy-2-mercaptopyrimidine, 4,6-Dihydroxypyrimidine-2-thiol, TBA
Empirical Formula (Hill Notation):
C
4
H
4
N
2
O
2
S
CAS Number:
504-17-6
Molecular Weight:
144.15
EC Number:
207-985-8
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Feature
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.08180
2-Thiobarbituric acid,
reagent for sorbic acid
Expand
View Pricing
T5500
2-Thiobarbituric acid,
≥98%
Expand
View Pricing