Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(2-Hydroxypropyl)-β-cyclodextrin
CAS Number:
128446-35-5
EC Number:
420-920-1
Products
Product Category
polysaccharides (3)
antibiotics (1)
media supplements (1)
Biological Source
Manufacturer
Melting Point (°C)
Physical Form
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
H107
(2-Hydroxypropyl)-β-cyclodextrin,
powder
Expand
View Pricing
332607
(2-Hydroxypropyl)-β-cyclodextrin,
average M
w
~1,460
Expand
View Pricing
332593
(2-Hydroxypropyl)-β-cyclodextrin,
average M
w
~1,380
Expand
View Pricing
778966
(2-Hydroxypropyl)-β-cyclodextrin,
Wacker Chemie AG
Expand
View Pricing
C0926
(2-Hydroxypropyl)-β-cyclodextrin,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
389145
(2-Hydroxypropyl)-β-cyclodextrin,
average M
w
~1,540
Expand
View Pricing
778907
(2-Hydroxypropyl)-β-cyclodextrin,
Wacker Chemie AG
Expand
View Pricing