
2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride

Empirical Formula (Hill Notation):
C9H12ClNO · HCl
CAS Number:
Molecular Weight:
222.11
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade