Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2′,7′-Dichlorofluorescein
Synonym(s):
2′,7′-Dichlorofluorescein
Empirical Formula (Hill Notation):
C
20
H
10
Cl
2
O
5
CAS Number:
76-54-0
Molecular Weight:
401.20
EC Number:
200-968-6
Products
Product Category
dyes (1)
fluorescence reagent (1)
stains (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
35848
2′,7′-Dichlorofluorescein,
BioReagent, suitable for fluorescence, ≥90% (T)
Expand
View Pricing
D6665
2′,7′-Dichlorofluorescein,
~90% purity (TLC) (85- 100% purity), powder
Expand
View Pricing
1.09676
2′,7′-Dichlorofluorescein,
fluorenscence indicator, ACS, Reag. Ph Eur
Expand
View Pricing
410217
2′,7′-Dichlorofluorescein,
ACS reagent
Expand
View Pricing