Skip to Content
Merck
2,4-Dichlorophenoxyacetic acid 97%

2,4-Dichlorophenoxyacetic acid

2,4-Dichlorophenoxyacetic acid 97%
Synonym(s):
2,4-D, 2,4-Dichlorophenoxyacetic acid
Linear Formula:
2,4-(Cl)2C6H3OCH2COOH
CAS Number:
Molecular Weight:
221.04

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing