Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,4,6-Tris(2-pyridyl)-
s
-triazine
Synonym(s):
2,4,6-Tri(2-pyridyl)-
s
-triazine, TPTZ
Empirical Formula (Hill Notation):
C
18
H
12
N
6
CAS Number:
3682-35-7
Molecular Weight:
312.33
EC Number:
222-965-9
Products
Brand
Supelco (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T1253
2,4,6-Tris(2-pyridyl)-
s
-triazine,
for spectrophotometric det. (of Fe), ≥98%
Expand
View Pricing
93285
2,4,6-Tris(2-pyridyl)-
s
-triazine,
for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Expand
View Pricing