Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,4,6-Trimethylpyridine
Synonym(s):
2,4,6-Collidine,
sym
-Collidine, 2,4,6-Trimethylpyridine
Linear Formula:
(CH
3
)
3
C
5
H
2
N
CAS Number:
108-75-8
Molecular Weight:
121.18
EC Number:
203-613-3
Products
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
142387
2,4,6-Trimethylpyridine,
ReagentPlus
®
, 99%
Expand
View Pricing
8.22267
2,4,6-Trimethylpyridine,
for synthesis
Expand
View Pricing
27690
2,4,6-Trimethylpyridine,
puriss. p.a., 99% (GC)
Expand
View Pricing