
20α-hydroxycholesterol

Synonym(s):
5-Cholestene-3β,20α-diol
Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
Products
Product Category
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing