
18-Crown-6

Synonym(s):
1,4,7,10,13,16-Hexaoxacyclooctadecane
Empirical Formula (Hill Notation):
C12H24O6
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing