Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,3-Dihydroxynaphthalene
Synonym(s):
1,3-Naphthalenediol, Naphthoresorcinol
Linear Formula:
C
10
H
6
(OH)
2
CAS Number:
132-86-5
Molecular Weight:
160.17
EC Number:
205-079-7
Products
Product Category
dyes (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
N6250
1,3-Dihydroxynaphthalene,
≥99% purity , crystalline
Expand
View Pricing
70650
1,3-Dihydroxynaphthalene,
for spectrophotometric det. of glucuronic acid according to Tollens, ≥97.0%
Expand
View Pricing