Skip to Content
Merck
All Photos(1)

Key Documents

1478301

USP

Olanzapine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Methyl-4-(4-methyl-1-piperazinyl)- 10H-thieno[2,3-b][1,5]benzodiazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4S
CAS Number:
Molecular Weight:
312.43
Beilstein:
7655141
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

olanzapine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24

InChI

1S/C17H20N4S/c1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12/h3-6,11,19H,7-10H2,1-2H3

InChI key

KVWDHTXUZHCGIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Olanzapine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Olanzapine and Fluoxetine Capsules
  • Olanzapine Orally Disintegrating Tablets
  • Olanzapine Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mauricio Tohen et al.
Archives of general psychiatry, 60(11), 1079-1088 (2003-11-12)
Despite the longer duration of the depressive phase in bipolar disorder and the frequent clinical use of antidepressants combined with antipsychotics or mood stabilizers, relatively few controlled studies have examined treatment strategies for bipolar depression. To examine the use of
Rajendra P Shukla et al.
Journal of neural transmission (Vienna, Austria : 1996), 127(2), 291-299 (2020-01-08)
Olanzapine is a thienobenzodiazepine compound. It is one of the newer types of antipsychotic drugs used in the treatment of schizophrenia and other psychotic disorders. Several methods have been reported for analyzing olanzapine in its pure form or combined with
Hirotake Hida et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(3), 601-613 (2014-08-15)
Blonanserin differs from currently used serotonin 5-HT₂A/dopamine-D₂ receptor antagonists in that it exhibits higher affinity for dopamine-D₂/₃ receptors than for serotonin 5-HT₂A receptors. We investigated the involvement of dopamine-D₃ receptors in the effects of blonanserin on cognitive impairment in an
Christopher M Hocking et al.
Supportive care in cancer : official journal of the Multinational Association of Supportive Care in Cancer, 22(4), 1143-1151 (2014-02-14)
Newer drugs incorporated in prophylactic regimens for chemotherapy-induced nausea and vomiting (CINV) have resulted in significantly reduced rates of this feared complication of cytotoxic chemotherapy. However, both delayed chemotherapy-induced nausea and breakthrough CINV remain difficult areas of management and require
Elizabeth Brunner et al.
BMC pharmacology & toxicology, 14, 38-38 (2013-08-02)
Olanzapine use has been reported during pregnancy and breastfeeding, but there are no controlled clinical trials assessing the safety of olanzapine exposure to infants and fetuses. The purpose of this report was to review and analyze prospective post-marketing cases of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service