Skip to Content
Merck
All Photos(1)

Key Documents

52999-U

Supelco

Supel Swift HLB DPX Tip

Universal format, Tip volume 1 mL, box of 96 tips, bed weight 20 mg

Synonym(s):

Supel Swift HLB DPX SPE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.21

product name

Supel Swift HLB DPX Tip Universal, weight 20 mg (bed), volume 1 mL, box of 96 tips

packaging

box of 96 tips

Quality Level

technique(s)

solid phase extraction (SPE): suitable

surface area

425-500 m2/g

volume

1 mL

weight

20 mg (bed)

matrix active group

Polymeric HLB phase

particle size

40-75 μm (>90%)

pore size

0.70-0.80 mL/g pore volume
80-210 Å pore size

compatibility

(MS-friendly)
Universal

Looking for similar products? Visit Product Comparison Guide

General description

DPX stands for Dispersive Pipette EXtraction. Supel Swift HLB DPX Tips are pipette tips that incorporate loosely contained Supel Swift HLB material, which is mixed with the sample solution to accomplish solid phase extraction in a dispersive manner. This DPX tip format allows for a simple and rapid workflow that is highly amenable to high throughput automation. Supel Swift HLB is a co-polymer solid phase extraction (SPE) phase composed of both hydrophilic and lipophilic functional groups. This phase is ideal for extracting a broad range of compounds from aqueous samples, including food/environmental samples as well as biological samples(eg. serum and plasma). Because of the hydrophilic and lipophilic balanced (HLB) property, the polymer material can selectively retain a wide range of compounds with different polarities and LogP values. Traditional HLB SPE uses a five-step process for sample preparation that normally includes the conditioning of sorbents with an organic solvent followed by an equilibration step to return sorbents to sample loading conditions. The product design allows for direct sample loading on to the sorbent bed, thereby eliminating the conditioning step altogether. This product feature can both save time and simplify sample preparation, which may also serve to reduce sample processing errors.

Application

  • Clinical
  • Bioanalysis
  • Pharmaceutical
  • Food & Bev
  • Environmental

Features and Benefits

  • Hydrophilic Modified Styrene Resin - extract and recover a broad range of analytes using a single sorbent (polar to nonpolar, acidic to basic)
  • Greater Capacity - allows for smaller bed weights, lower elution volumes, and time savings in sample processing
  • Resistant to Over-Drying - robust methodology, lower variability
  • Stringent Production & QC Standards - greater product consistency for improved accuracy and precision over all SPE formats

Analysis Note

The 96-tip dispersive SPE format is perfect for medium to high-throughput laboratories performing large numbers of samples at a time as the format is automation compatible. Recommended weight tip based on sample volume:

  • Sample Volume: <300 µL (Bed Weight: 3 mg or 5 mg)
  • Sample Volume: <600 µL (Bed Weight: 10 mg)
  • Sample Volume: <800 µL (Bed Weight: 20 mg)

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service