Skip to Content
Merck
All Photos(1)

Documents

442687

Supelco

N-Nitrosodiethylamine

analytical standard

Synonym(s):

Diethylnitrosamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NNO
CAS Number:
Molecular Weight:
102.14
Beilstein:
1744991
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

177 °C (lit.)

density

0.95 g/mL (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CCN(CC)N=O

InChI

1S/C4H10N2O/c1-3-6(4-2)5-7/h3-4H2,1-2H3

InChI key

WBNQDOYYEUMPFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Verna et al.
Pharmacology & therapeutics, 71(1-2), 57-81 (1996-01-01)
N-Nitrosodiethylamine (NDEA) is DNA reactive after bioactivation and produces tumors in every animal species tested. Bioactivation is effected by several P450 isozymes including CYP2E1, which is ethanol inducible. Tumor formation in rat liver was proportional to O4-ethyldeoxythymidine formation in DNA
Takeki Uehara et al.
Toxicological sciences : an official journal of the Society of Toxicology, 132(1), 53-63 (2013-01-05)
Hepatocellular carcinoma (HCC) mostly develops in patients with advanced fibrosis; however, the mechanisms of interaction between a genotoxic insult and fibrogenesis are not well understood. This study tested a hypothesis that fibrosis promotes HCC via a mechanism that involves activation
Ziyan Wang et al.
Hepatology (Baltimore, Md.), 57(5), 1869-1881 (2013-01-10)
Hepatocellular carcinoma (HCC) is a devastating consequence of chronic inflammatory liver diseases. The goal of this study was to investigate whether Toll-like receptor 4 (TLR4) activity contributes to HCC initiation and progression in mice. A mouse model of diethylnitrosamine (DEN)-induced
Jingling Jin et al.
The Journal of biological chemistry, 288(20), 14451-14462 (2013-04-09)
Cancer changes biological processes in the liver by altering gene expression at the levels of transcription, translation, and protein modification. The RNA binding protein CUGBP1 is a key regulator of translation of CCAAT enhancer binding protein β and histone deacetylase
Marta Anna Kowalik et al.
Oncotarget, 6(36), 38749-38763 (2015-10-10)
Although the expression of the stem/progenitor cell marker cytokeratin-19 (CK-19) has been associated with the worst clinical prognosis among all HCC subclasses, it is yet unknown whether its presence in HCC is the result of clonal expansion of hepatic progenitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service