Skip to Content
Merck
All Photos(1)

Key Documents

13019AST

Supelco

Astec® CHIROBIOTIC® R Chiral (5 μm) HPLC Columns

L × I.D. 15 cm × 2.1 mm, HPLC Column

Synonym(s):

CHIROBIOTIC R Chiral Chromatography Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm

material

stainless steel column

description

HPLC column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

15 cm × 2.1 mm

matrix

high-purity silica gel particle platform
fully porous particle

matrix active group

ristocetin A glycopeptide phase

particle size

5 μm

pore size

100 Å

operating pH range

3.5-6.8

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

CHIROBIOTIC® R, based on the ristocetin A glycopeptide covalently bonded to high purity silica particles, has shown particular applicability to enantiomers of anionic compounds. Selectivity on CHIROBIOTIC® R strongly correlates to the organic modifier, favoring the alcohol-type mobile phases by a large margin.

  • Bonded phase: Ristocetin A
  • Operating pH range: 3.5 - 6.8
  • Particle diameter: 5, 10 or 16 μm
  • Pore size: 100 Å

CHIROBIOTIC FAQs
CHIROBIOTIC Reference Bibliography
Chiral Product Literature

Application


  • Sensitive determination of D-lactic acid and L-lactic acid in urine by high-performance liquid chromatography-tandem mass spectrometry.: This study demonstrates the application of the Astec CHIROBIOTIC R Chiral HPLC Column in the sensitive determination of D-lactic acid and L-lactic acid in urine samples using HPLC-MS/MS. This highlights its utility in clinical and biomedical research for the analysis of chiral molecules in biological fluids (Henry et al., 2012).

  • New high-performance liquid chromatography method for the determination of (R)-warfarin and (S)-warfarin using chiral separation on a glycopeptide-based stationary phase.: This research utilizes the Astec CHIROBIOTIC R Chiral HPLC Column to develop a method for the chiral separation and determination of warfarin enantiomers. It underscores the column′s application in pharmaceutical analysis, particularly in the study of chiral drug enantiomers (Malakova et al., 2009).



Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzyme-catalyzed synthesis of (R)- and (S)-3-hydroxy-3-(10-alkyl-10H-phenothiazin-3-yl)propanoic acids
Brem, Jurgen, et al.
Tetrahedron Asymmetry, 21 (3), 365-373 (2010)
P Dehouck et al.
Journal of chromatography. A, 1010(1), 63-74 (2003-09-25)
Erythromycin is a mixture of macrolide antibiotics produced by Saccharopolyspora erythreas during fermentation. A new method for the analysis of erythromycin by liquid chromatography has previously been developed. It makes use of an Astec C18 polymeric column. After validation in
H Henry et al.
Biomedical chromatography : BMC, 26(4), 425-428 (2011-08-16)
D-lactic acid in urine originates mainly from bacterial production in the intestinal tract. Increased D-lactate excretion as observed in patients affected by short bowel syndrome or necrotizing enterocolitis reflects D-lactic overproduction. Therefore, there is a need for a reliable and
B DeCruze et al.
Clinical oncology (Royal College of Radiologists (Great Britain)), 11(4), 252-254 (1999-09-04)
Poor prognosis (poorly differentiated and/or deep myometrial invasion) Stage I endometrial cancer can have a relapse rate as high as 50%. Traditionally, most clinical oncologists treat these patients with external beam radiotherapy after surgery but there is no evidence to
V Wsól et al.
Journal of chromatography. B, Biomedical sciences and applications, 689(1), 205-214 (1997-02-07)
The major metabolite of a novel non-steroidal anti-inflammatory drug, DL-4-(2',4'-difluorobiphenyl-4-yl)-2-oxo-2-methylbutanoic acid (flobufen, I), namely 4-(2',4'-difluorobiphenyl-4-yl)-2-methyl-gamma-butyrolactone (4-dihydroflobufen lactone, III), has four stereoisomers consisting of two racemic pairs of enantiomers. Of three chiral stationary phases tested, Cyclobond I beta-RSP (Astec) (beta-cylodextrin derivatized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service