Skip to Content
Merck
All Photos(7)

Key Documents

ZRB1621

Sigma-Aldrich

Anti-phospho-ULK1-Ser757 Antibody, clone 5L18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ATG1, Autophagy-related protein 1 homolog, EC:2.7.11.1, Serine/threonine-protein kinase ULK1, Unc-51-like kinase 1, hATG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5L18, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 112.63 kDa
observed mol wt ~112 kDa

species reactivity

human

species reactivity (predicted by homology)

sheep, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer75)

Gene Information

human ... ULK1(8408)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 5L18 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects ULK1 phosphorylated on serine 757.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids surrounding phosphoserine 757 from the C-terminal half of human ULK1.

Application

Anti-phospho-ULK1-Ser757, clone 5L18 ZooMAb, Cat. No. ZRB1621, is a recombinant Rabbit monoclonal antibody that targets ULK1 phosphorylated on serine 757 and is tested in Affinity Binding Assay and Peptide Inhibition Assay, and Western Blotting.
Peptide Inhibition Assay: Target band detection in lysate from serum starved A431 cells was prevented by preblocking of a representative lot (1:1,000 dilution) with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Affinity Binding Assay: A representative lot of this antibody bound phosphorylated ULK1 (pSer757) peptide with a KD of 8.2 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Serine/threonine-protein kinase ULK1 (UniProt: O75385; also known as EC:2.7.11.1, Autophagy-related protein 1 homolog, ATG1, hATG1, Unc-51-like kinase 1) is encoded by the ULK1 (also known as KIAA0722) gene (Gene ID: 8408) in human. ULK1 is a ubiquitously expressed serine-threonine protein kinase that is involved in autophagy in response to starvation. During starvation it is localized to puncate structures primarily representing the isolation membrane that sequesters a portion of the cytoplasm resulting in the formation of an autophagosome. In cells, ULK1 is constitutively in complex with at least three proteins: ATG13, FIP200 and ATG101. Interaction of ULK1 with ATG13 or FIP200 is reported to increase ULK1 kinase activity and stability. ULK1 undergoes autophosphorylation under nutrient-rich conditions and is dephosphorylated during starvation or following treatment with rapamycin. Under nutrient sufficiency, it is phosphorylated by MTOR/mTOR, which disrupts the interaction with AMPK and prevents activation of ULK1. In response to nutrient limitation it is phosphorylated and activated by AMPK that leads to activation of autophagy. It can also act as a regulator of AMPK by mediating phosphorylation of AMPK subunits PRKAA1, PRKAB2 and PRKAG1, leading to negatively regulation of AMPK activity. Under nutrient sufficiency, high mTOR activity is reported to prevent ULK1 activation by phosphorylating ULK1 on serine 757 and disrupt its interaction with AMPK. The protein kinase domain of ULK1 is localized to amino acids 16-278. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Zachari, M., and Ganley, IG. (2017). Essays Biochem. 61(6); 585-596; Kim, J., et al. (2011). Nat. Cell Biol. 13(2); 132-141).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service