Skip to Content
Merck
All Photos(4)

Documents

X2502

Sigma-Aldrich

Xanthine sodium salt

≥99%

Synonym(s):

2,6-Dihydroxypurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3N4NaO2
CAS Number:
Molecular Weight:
174.09
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

powder

impurities

≤4% total solvent

solubility

dilute NaOH: soluble, clear to slightly hazy

SMILES string

[Na].O=C1NC(=O)c2[nH]cnc2N1

InChI

1S/C5H4N4O2.Na.H/c10-4-2-3(7-1-6-2)8-5(11)9-4;;/h1H,(H3,6,7,8,9,10,11);;

InChI key

JAZQMXVZANZJSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xanthine is an intermediate of purine nucleotide metabolism. It is found in most body tissues and fluids. Xanthine derivatives are present in coffee, tea, and chocolate.

Application

Xanthine sodium salt has been used as a component of substrate solution for xanthine oxidase during superoxide dismutase (SOD) assay. It has also been used as a supplement in a special-conditioned medium to culture Swiss 3T3 fibroblasts stably transfected with tetracycline (tet)-controlled transactivator and transactivator-controlled tissue transglutaminase cDNA [39] constructs (clone TG3).

Biochem/physiol Actions

Xanthine acts as a substrate for xanthine oxidase. It elevates cellular cyclic AMP levels by preventing its breakdown and metabolism by inhibition of tissue phosphodiesterases. Xanthine also exhibits anti-inflammatory properties either by the release of anti-inflammatory cytokines or gene transcription regulation or histone deacetylase activation. All these properties of xanthine aid in the relaxation of smooth muscles of the bronchial tree.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

İnci Kurt-Celep et al.
Matrix biology : journal of the International Society for Matrix Biology, 105, 1-16 (2021-11-12)
In wound healing, the TG2 enzyme plays a dual functional role. TG2 has been shown to regulate extracellular matrix (ECM) stabilization by its transamidase activity while increasing cell migration by acting as a cell adhesion molecule. In this process, nitric
Munevver B Cicekdal et al.
IUBMB life, 71(12), 1973-1985 (2019-08-20)
Calorie restriction (CR) is one of the most effective methods to prevent many diseases including cancer in preclinical models. However, the molecular mechanism of how CR prevents cancer is unclear. The aim of this study was to understand the role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service