Skip to Content
Merck
All Photos(1)

Key Documents

T2952

Sigma-Aldrich

Tetrabenazine

≥98% (HPLC), solid

Synonym(s):

9,10-Dimethoxy-1,3,4,6,7,11b-hexahydro-3-isobutyl-(rel 3R,11bR)-2H-benzo[a]quinolizin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27NO3
CAS Number:
Molecular Weight:
317.42
Beilstein:
40090
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL
H2O: insoluble

originator

Roche

storage temp.

2-8°C

SMILES string

COc1cc2CCN3C[C@@H](CC(C)C)C(=O)CC3c2cc1OC

InChI

1S/C19H27NO3/c1-12(2)7-14-11-20-6-5-13-8-18(22-3)19(23-4)9-15(13)16(20)10-17(14)21/h8-9,12,14,16H,5-7,10-11H2,1-4H3/t14-,16-/m1/s1

InChI key

MKJIEFSOBYUXJB-GDBMZVCRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetrabenazine has been used for dopamine uptake assays in mouse brain cells1. Tetrabenazine has also been used for non-specific binding assays in postnuclear supernatants derived from PC-12 and CV-1 cells2.

Biochem/physiol Actions

Tetrabenazine is a reversible type 2 vesicular monoamine transporter (VMAT) inhibitor.
Tetrabenazine is a reversible type 2 vesicular monoamine transporter (VMAT) inhibitor. It depletes dopamine stores.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Tetrabenazine is soluble in DMSO at a concentration that is greater than 10 mg/ml and is insoluble in water.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael P Dalton et al.
eLife, 12 (2024-03-22)
The vesicular monoamine transporter 2 (VMAT2) is a proton-dependent antiporter responsible for loading monoamine neurotransmitters into synaptic vesicles. Dysregulation of VMAT2 can lead to several neuropsychiatric disorders including Parkinson's disease and schizophrenia. Furthermore, drugs such as amphetamine and MDMA are
Mahabuba Jahan et al.
EJNMMI research, 1(1), 33-33 (2012-01-05)
Fluorine-18 dihydrotetrabenazine [DTBZ] analogues, which selectively target the vesicular monoamine transporter 2 [VMAT2], have been extensively studied for in vivo quantification of beta cell mass by positron-emission tomography [PET]. This study describes a novel deuterated radioligand [18F]fluoroethyl [FE]-DTBZ-d4, aimed to
Marc D Normandin et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 53(6), 908-916 (2012-05-11)
The ability to noninvasively measure endogenous pancreatic β-cell mass (BCM) would accelerate research on the pathophysiology of diabetes and revolutionize the preclinical development of new treatments, the clinical assessment of therapeutic efficacy, and the early diagnosis and subsequent monitoring of
Sinéad M Murphy et al.
Annals of neurology, 72(4), 481-490 (2012-10-31)
Since the introduction of the Orphan Drug Act in 1983, designed to promote development of treatments for rare diseases, at least 378 orphan drugs have been approved. Incentives include financial support, tax credits, and perhaps most importantly, extended market exclusivity.
Nicolaas I Bohnen et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 32(8), 1609-1617 (2012-05-10)
Parkinson's disease (PD) is a multisystem neurodegenerative disorder. Heterogeneous clinical features may reflect heterogeneous changes in different brain regions. In contrast to the pronounced nigrostriatal denervation characteristic of PD, cholinergic changes are less marked. We investigated cholinergic innervation activity in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service