Skip to Content
Merck
All Photos(2)

Documents

T2402

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−TRITC antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Rabbit Anti-Mouse IgG (whole molecule)−Tetramethylrhodamine isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

TRITC conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:64
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoglobulins (Igs) belongs immunoglobulin super-family. There are five different types of Immunoglobulins. Each immunoglobulin has two heavy (H) and two light (L), held together by disulphide linkages. Each light comprises one variable N-terminal region and a constant C-terminal region. Heavy chain has one variable N-terminal region and three or four constant (CH1-CH4) C-terminal region. IgG is abundant immunoglobulin in human serum. The four classes of IgG can be IgG1, IgG2, IgG3, and IgG4.

Specificity

Anti-Mouse IgG (whole molecule)-TRITC antibody is specific for mouse IgG.

Immunogen

Normal mouse serum

Application

Anti-Mouse IgG (whole molecule)-TRITC antibody is suitable for use in direct immunofluorescence (1:64), immunohistochemistry (1:200 dilution in formalin-fixed, paraffin-embedded sections), Ouchterlony double diffusion and electrophoresis techniques.

Biochem/physiol Actions

Digestion of IgG by papain results in generation of fragment antigen binding (Fab) comprising one complete L chain and a variable and CH1 region of H chain. Pepsin digestion of IgG results in fragment crystallizable (fc), comprises the H chain constant region.
Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice .

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica E Poort et al.
Human molecular genetics, 25(17), 3768-3783 (2016-08-06)
Spinal bulbar muscular atrophy (SBMA) is a progressive, late onset neuromuscular disease causing motor dysfunction in men. While the morphology of the neuromuscular junction (NMJ) is typically affected by neuromuscular disease, whether NMJs in SBMA are similarly affected by disease
Rajbir K Gill et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 14(22), 7481-7487 (2008-11-18)
We seek to establish a genetic test to identify lung cancer using cells obtained through computed tomography-guided fine needle aspiration (FNA). We selected regions of frequent copy number gains in chromosomes 1q32, 3q26, 5p15, and 8q24 in non-small cell lung
G Mozes et al.
Arteriosclerosis, thrombosis, and vascular biology, 18(7), 1157-1163 (1998-07-22)
Macrophage colony stimulating factor (MCSF) is believed to play a key role in one of the earliest events in atherosclerosis, ie, monocyte to macrophage differentiation in the arterial intima. The aim of this study was to examine the biological effects
Su-Jin Heo et al.
eLife, 5 (2016-12-03)
Mesenchymal stem cell (MSC) differentiation is mediated by soluble and physical cues. In this study, we investigated differentiation-induced transformations in MSC cellular and nuclear biophysical properties and queried their role in mechanosensation. Our data show that nuclei in differentiated bovine
X A Bigard et al.
Journal of applied physiology (Bethesda, Md. : 1985), 81(6), 2658-2665 (1996-12-01)
The aim of this study was to analyze the effects of treadmill training (2 h/day, 5 days/wk, 30 m/min, 7% grade for 5 wk) on the expression of myosin heavy chain (MHC) isoforms during and after regeneration of a fast-twitch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service