Skip to Content
Merck
All Photos(1)

Key Documents

SRP3309

Sigma-Aldrich

GDNF human

recombinant, expressed in E. coli, Animal-component free, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

ATF-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.1 μg/mL

mol wt

30.0 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GDNF(2668)

General description

Glial cell derived neurotrophic factor (GDNF) is a disulfide-linked homodimeric neurotrophic factor structurally related to Artemin, Neurturin and Persephin. These proteins belong to the cysteine-knot superfamily of growth factors that assume stable dimeric protein structures. The gene encoding this protein is localized on human chromosome 5p13.2. The functional human GDNF ligand is a disulfide-linked homodimer, of two 15kDa polypeptide chains called monomers. Each monomer contains seven conserved cysteine residues, one of which (Cys 101) is used for inter-chain disulfide bridging and the others are involved in intramolecular ring formation known as the cysteine knot configuration.

Application

Glial cell derived neurotrophic factor (GDNF) has been used as a supplement in dorsal root ganglion (DRG) culture medium to study the effect of growth factors on neuron viability, neurite outgrowth and arborisation. (3)

Biochem/physiol Actions

Glial cell derived neurotrophic factor (GDNF) signals through a multicomponent receptor system, composed of a RET and one of the four GFR-α (α1-4) receptors. GDNF specifically promotes dopamine uptake and survival and morphological differentiation of midbrain neurons. Using Parkinson′s disease mouse model, GDNF has been shown to improve conditions such as bradykinesia, rigidity, and postural instability.

Physical form

Lyophilized from 10 mM Sodium Citrate + 50 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enrique Pimentel
Handbook of Growth Factors, Volume 2 null
Primary Postnatal Dorsal Root Ganglion Culture from Conventionally Slaughtered Calves.
Fadda A
PLoS ONE, 11(12), e0168228-e0168228 (2016)
Jacob L Roam et al.
Biomaterials, 35(24), 6473-6481 (2014-05-13)
Introduction of spatial patterning of proteins, while retaining activity and releasability, is critical for the field of regenerative medicine. Reversible binding to heparin, which many biological molecules exhibit, is one potential pathway to achieve this goal. We have covalently bound
Analysis of RET, ZEB2, EDN3 and GDNF genomic rearrangements in 80 patients with Hirschsprung disease (using multiplex ligation-dependent probe amplification).
Serra A
Annals of Human Genetics, 73(2), 147-151 (2009)
L F Lin et al.
Science (New York, N.Y.), 260(5111), 1130-1132 (1993-05-21)
A potent neurotrophic factor that enhances survival of midbrain dopaminergic neurons was purified and cloned. Glial cell line-derived neurotrophic factor (GDNF) is a glycosylated, disulfide-bonded homodimer that is a distantly related member of the transforming growth factor-beta superfamily. In embryonic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service