Skip to Content
Merck
All Photos(1)

Key Documents

SML2622

Sigma-Aldrich

ABC1183

≥98% (HPLC)

Synonym(s):

4-(4-Amino-2-p-tolylamino-thiazole-5-carbonyl)-benzonitrile, 4-[[4-Amino-2-[(4-methylphenyl)amino]-5-thiazolyl]carbonyl]-benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N4OS
CAS Number:
Molecular Weight:
334.39
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=C(C(C2=CC=C(C=C2)C#N)=O)SC(NC3=CC=C(C)C=C3)=N1

Biochem/physiol Actions

ABC1183 is an orally active, potent and selective inhibitor of GSK3 α/β and CDK9, which induces G2/M cell cycle arrest and apoptosis in cancer cells. Apparently ABC1183 suppresses tumor growth and inflammation-driven gastrointestinal disease symptoms due to down regulation of TNFα and IL-6 pro-inflammatory cytokines. It does not exhibit organ or hematological toxicities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Randy S Schrecengost et al.
The Journal of pharmacology and experimental therapeutics, 365(1), 107-116 (2018-02-13)
Glycogen synthase kinase-3s (GSK3α and GSK3β) are constitutively active protein kinases that target over 100 substrates, incorporate into numerous protein complexes, and regulate such vital cellular functions as proliferation, apoptosis, and inflammation. Cyclin-dependent kinase 9 (CDK9) regulates RNA production as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service