Skip to Content
Merck
All Photos(1)

Key Documents

SML1643

Sigma-Aldrich

AS2444697

≥98% (HPLC)

Synonym(s):

AS 2444697, N-[3-(Aminocarbonyl)-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-yl-2-(2-methyl-4-pyridinyl)-4-Oxazolecarboxamide hydrochloride (1:1), N-[3-Carbamoyl-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-yl]-2-(2-methylpyridin-4-yl)-1,3-oxazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N6O4 · HCl
CAS Number:
Molecular Weight:
432.86
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(C1=COC(C2=CC=NC(C)=C2)=N1)NC3=CN(C4CCOCC4)N=C3C(N)=O.[H]Cl

InChI

1S/C19H20N6O4.ClH/c1-11-8-12(2-5-21-11)19-23-15(10-29-19)18(27)22-14-9-25(24-16(14)17(20)26)13-3-6-28-7-4-13;/h2,5,8-10,13H,3-4,6-7H2,1H3,(H2,20,26)(H,22,27);1H

InChI key

FGNHLIIFEDYNFZ-UHFFFAOYSA-N

Application

AS2444697 has been used as an interleukin-1R-associated kinase 4 (IRAK4) inhibitor to study the role of interleukin (IL)-1/IL-33 signaling in the development of endometriosis using a mouse model of endometriosis. It has also been used to study its dose-dependent effect on the C-X-C motif chemokine ligand 8 (CXCL8) gene expression

Biochem/physiol Actions

AS2444697 is a potent and selective inhibitor of interleukin-1 receptor-associated kinase 4 (IRAK-4) that suppresses the progression of chronic renal failure via anti-inflammatory action.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Germline gain-of-function myeloid differentiation primary response gene-88 (MYD88) mutation in a child with severe arthritis.
Keith A Sikora et al.
The Journal of allergy and clinical immunology, 141(5), 1943-1947 (2018-02-11)
Toru Kato et al.
Frontiers in immunology, 10, 2021-2021 (2019-09-12)
Endometriosis is an estrogen-dependent disease with symptoms of dysmenorrhea, chronic pain, and infertility that affects 6-10% of women of reproductive age. Medical or surgical therapy, such as administration of an anti-gonadotropin or ovarian cystectomy, provide effective pain relief. However, neither

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service