Skip to Content
Merck
All Photos(1)

Key Documents

SML0870

Sigma-Aldrich

LY 2087101

≥98% (HPLC)

Synonym(s):

(2-Amino-5-keto)thiazole), [2-[(4-Fluorophenyl)amino]-4-methyl-5-thiazolyl]-3-thienyl-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11FN2OS2
CAS Number:
Molecular Weight:
318.39
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

(dry room)

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C15H11FN2OS2/c1-9-14(13(19)10-6-7-20-8-10)21-15(17-9)18-12-4-2-11(16)3-5-12/h2-8H,1H3,(H,17,18)

InChI key

PEAMDZVDNYENPN-UHFFFAOYSA-N

Biochem/physiol Actions

LY 2087101 is a selective positive allosteric potentiator of α7 and α4β2 nicotinic acetylcholine receptors (nAChRs). LY 2087101 was shown to not enhance the activity of α3β4 subtype nAChRs.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service