Skip to Content
Merck
All Photos(1)

Key Documents

SML0744

Sigma-Aldrich

LDN-27219

≥98% (HPLC)

Synonym(s):

2-[(3,4-Dihydro-4-oxo-3,5-diphenylthieno[2,3-d]pyrimidin-2-yl)thio]-acetic acid hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16N4O2S2
CAS Number:
Molecular Weight:
408.50
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

InChI

1S/C20H16N4O2S2/c21-23-16(25)12-28-20-22-18-17(15(11-27-18)13-7-3-1-4-8-13)19(26)24(20)14-9-5-2-6-10-14/h1-11H,12,21H2,(H,23,25)

InChI key

WLBUICQBNZXIDJ-UHFFFAOYSA-N

Biochem/physiol Actions

LDN-27219 is a revesible inhibitor of transglutaminase 2 (TG2) that binds to the GTP binidng site of the enzyme and inhbits activity with and IC50 of 600 nM.
LDN-27219 is a thieno[2,3-d]pyrimidine-4-one-acylhydrazide derivative. Closed conformation of TG2 induced by LDN-27219 can be a potential therapeutic strategy for age-related vascular dysfunction and lowering blood pressure.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estéfano Pinilla et al.
Hypertension (Dallas, Tex. : 1979), 77(1), 216-227 (2020-12-01)
Transglutaminase 2 (TG2) is an enzyme which in the open conformation exerts transamidase activity, leading to protein cross-linking and fibrosis. In the closed conformation, TG2 participates in transmembrane signaling as a G protein. The unspecific transglutaminase inhibitor cystamine causes vasorelaxation
Nayeon Kim et al.
Organic & biomolecular chemistry, 12(27), 4932-4940 (2014-06-01)
In the process of optimization, we developed a novel core skeleton of thieno[3,4-b]pyrazine via GK-13. The derivatives synthesized were shown to inhibit TGase 2 activity in cancer cells. Some of the hit compounds such as the arylethynyl group-coupled thieno[3,4-b]pyrazine derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service