Skip to Content
Merck
All Photos(1)

Documents

SML0330

Sigma-Aldrich

S3I-201

≥97% (HPLC)

Synonym(s):

2-Hydroxy-4-[[[[(4-methylphenyl)sulfonyl]oxy]acetyl]amino]-benzoic acid, NSC 74859

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15NO7S
CAS Number:
Molecular Weight:
365.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCC(=O)Nc2ccc(C(O)=O)c(O)c2

InChI

1S/C16H15NO7S/c1-10-2-5-12(6-3-10)25(22,23)24-9-15(19)17-11-4-7-13(16(20)21)14(18)8-11/h2-8,18H,9H2,1H3,(H,17,19)(H,20,21)

InChI key

HWNUSGNZBAISFM-UHFFFAOYSA-N

Application

S3I-201 has been used as a signal transducer and activator of transcription 3 (STAT3) inhibitor:
  • to confirm the role of STAT3 phosphorylation in interleukin (IL)-33 production in lung epithelial cells and IL-22 mRNA expression in sorted group 3 innate lymphoid cells (ILC3s)
  • to study the cellular response of STAT3 triggered by β-hexaclorocyclohexane (β-HCH) in various cell lines
  • to examine the influence of STAT3 in response to angiotensin II (ang II) on induction of fibrotic proteins in kidney epithelial cells

Biochem/physiol Actions

Inhibition of signal transducer and activator of transcription 3 (STAT3) activity by S3I-201 may be a potential therapeutic strategy for hypertensive kidney disease.
S3I-201 is a cell-permeable Stat3 inhibitor that binds to the Stat3-SH2 domain, prevents Stat3 phosphorylation/activation, dimerization, and DNA-binding.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safiye E Sarper et al.
Scientific reports, 8(1), 10906-10906 (2018-07-22)
Rodent incisors grow permanently and the homeostasis of enamel production is maintained by a continuous supply of epithelial progenitors from putative stem cells in the cervical loop. We herein report that Runx1 regulates the Lgr5-expressing epithelial stem cells and their
Sang-Hun Kim et al.
Molecular medicine reports, 16(6), 9224-9232 (2017-10-11)
Chlorogenic acid (CA) is a phenolic compound purified from coffee, fruits and their associated beverages, which possess various biological properties, such as antioxidant and anticarcinogenic activities. The present study evaluated the effects of CA on lipopolysaccharide (LPS)‑induced inflammation in RAW264.7
Wei-Jan Wang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(2), 503-513 (2016-07-21)
Cisplatin (CDDP) is frequently used in combination chemotherapy with paclitaxel for treating urothelial carcinoma of the urinary bladder (UCUB). CDDP cross-resistance has been suggested to develop with paclitaxel, thus hindering successful UCUB treatment. Therefore, elucidating the mechanisms underlying CDDP-induced anticancer
Kwang Bo Jung et al.
Journal of clinical medicine, 8(7) (2019-07-07)
Human intestinal organoids (hIOs), which resemble the human intestine structurally and physiologically, have emerged as a new modality for the study of the molecular and cellular biology of the intestine in vitro. We recently developed an in vitro maturation technique
Gaurav Kumar et al.
Radiology, 286(2), 524-536 (2017-09-08)
Purpose To (a) identify key expressed genes in the periablational rim after radiofrequency ablation (RFA) and their role in driving the stimulation of distant tumor growth and (b) use adjuvant drug therapies to block key identified mediator(s) to suppress off-target

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service