Skip to Content
Merck
All Photos(1)

Documents

SML0320

Sigma-Aldrich

FR180204

≥98% (HPLC)

Synonym(s):

5-(2-Phenyl-pyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-c]pyridazin-3-ylamine, 5-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-c]pyridazin-3-amine, FR 180204

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13N7
CAS Number:
Molecular Weight:
327.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

faintly yellow to dark yellow

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

Nc1n[nH]c2nnc(cc12)-c3c(nn4ccccc34)-c5ccccc5

InChI

1S/C18H13N7/c19-17-12-10-13(20-22-18(12)23-21-17)15-14-8-4-5-9-25(14)24-16(15)11-6-2-1-3-7-11/h1-10H,(H3,19,21,22,23)

InChI key

XVECMUKVOMUNLE-UHFFFAOYSA-N

Application

FR180204 has been used as ERK inhibitor:
  • to block extracellular-signal-regulated kinase (ERK) in order to validate whether homeobox B7 (HOXB7) regulates the migration and proliferation process via AKT/mitogen-activated protein kinases (MAPK) signaling
  • of extracellular-signal-regulated kinase (ERK) to study the effects of signaling pathway inhibitors on differentiation and cell traction stress
  • to determine if phosphorylation of this signaling protein is essential for mesenchymal stromal cell (MSC) derived therapeutic efficacy

Biochem/physiol Actions

FR180204 is a potent, cell-permeable, ATP-competitive inhibitor of ERK1 and ERK2 (mitogen-activated protein kinase (MAPK)/extracellular-signal-regulated kinases (ERK) 1/2).
It can be used to study the roles of ERK as well as for drug development.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Substrate stiffness and matrix composition coordinately control the differentiation of liver progenitor cells
Kourouklis AP, et al.
Biomaterials, 99, 82-94 (2016)
Mario Sechi et al.
Scientific reports, 8(1), 15726-15726 (2018-10-26)
The anti-proliferative activity of dietary flavonoid fisetin has been validated in various cancer models. Establishing its precise mechanism of action has proved somewhat challenging given the multiplicity of its targets. We demonstrated that YB-1 promotes epithelial-to-mesenchymal transition and its inhibition
Miho Tanaka et al.
Cancer research, 78(13), 3589-3603 (2018-04-25)
Natural killer (NK) cells express receptors specific for MHC class I (MHC-I) molecules involved in "missing-self" recognition of cancer and virus-infected cells. Here we elucidate the role of MHC-I-independent NKR-P1B:Clr-b interactions in the detection of oncogenic transformation by NK cells.
Identification of a selective ERK inhibitor and structural determination of the inhibitor-ERK2 complex
Ohori M, et al.
Biochemical and biophysical research communications, 336(1), 357-363 (2005)
Maren Bakkebø et al.
BMC immunology, 11, 57-57 (2010-11-26)
Cytokines of the transforming growth factor β (TGF-β) superfamily exert effects on proliferation, apoptosis and differentiation in various cell types. Cancer cells frequently acquire resistance to the anti-proliferative signals of TGF-β, which can be due to mutations in proteins of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service