Skip to Content
Merck
All Photos(1)

Key Documents

SMB01024

Sigma-Aldrich

Diuvaretin

≥90% (LC/MS-ELSD)

Synonym(s):

1-[2,4-dihydroxy-3,5-bis[(2-hydroxyphenyl)methyl]-6-methoxyphenyl]-3-phenylpropan-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H28O6
CAS Number:
Molecular Weight:
484.54
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

484.54

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C30H28O6/c1-36-30-23(18-21-12-6-8-14-25(21)32)28(34)22(17-20-11-5-7-13-24(20)31)29(35)27(30)26(33)16-15-19-9-3-2-4-10-19/h2-14,31-32,34-35H,15-18H2,1H3

InChI key

KDEMELRYKGOXDL-UHFFFAOYSA-N

General description

Diuvaretin, a dihydrochalcone, is a natural product commonly available from Dipteryx odorata, Uvaria acuminata and Uvaria chamae plants. This plant metabolite exhibits various biological activities such as anti-inflammatory, antioxidant, antineoplastic, anti-angiogenic, and antimalarial properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Diuvaretin exhibits cytotoxicity towards human promyelocytic leukemia HL-60 cells. It has a role as a metabolite, an antineoplastic agent and an antimalarial.
Diuvaretin inhibits NF-kB activation, hindering inflammation and cancer. It also suppresses tumor cell proliferation, induces apoptosis, and diminishes tumor growth and angiogenesis in mouse cancer models.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service