Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501174

Sigma-Aldrich

Anti-Connexin 43 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CX43, CXA1, CXN-43, Connexin 43, Gap junction 43 kDa heart protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 43 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GJA1(2697)

Related Categories

General description

Connexin 43 belongs to the connexin protein family that forms gap junction. This protein is highly expressed in neurons and radial glia during development and in astrocytes in adult brains.
Connexin 43 belongs to the connexin protein family that forms gap junction. This protein is highly expressed in neurons and radial glia during development and in astrocytes in adult brains.

Immunogen

The antiserum was produced against synthesized peptide derived from human Connexin 43.

Immunogen Range: 332-381

Application

Anti-Connexin 43, C-Terminal antibody produced in rabbit has been used in flow cytometry.

Biochem/physiol Actions

In rodents, connexin-43 (Cx-43) shows specific temporal patterns of expression during brain development. Experimental studies reveal that, connexin-43 expression increases in the brain during sheep ontogenesis.This protein might play a crucial role in neuronal migration as it resides in radial glial fibers which are closely associated with migrating neurons. It is also involved in transmembrane NAD+ transport. connexin-43, which is an element of gap junction, helps in cell–cell communication. Cx-43 is known to induce [Ca2+] i burst phenomenon in uterine artery endothelial cells (UAECs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grazyna B Sadowska et al.
Brain research, 1255, 51-56 (2008-12-23)
Gap junctions are specialized membrane structures that mediate intercellular communication and facilitate passage of ions and small molecules between adjacent cells. Connexins comprise a multigene family of transmembrane proteins that form gap junctions. Connexin-32 and connexin-43 are among the most
B Nadarajah et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 17(9), 3096-3111 (1997-05-01)
Gap junctions are membrane channels that mediate the direct passage of ions and molecules between adjacent cells. Recent tracer coupling and optical recording studies have revealed the presence of gap junction-mediated communication between neurons during neocortical development. We have visualized
S Bruzzone et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 15(1), 10-12 (2000-12-02)
A previously unrecognized passive transport for pyridine dinucleotides has been described recently in the plasmamembrane of several mammalian cells. Despite elucidation of some functional and kinetic properties of this transport system, it is still undefined at the molecular level. Therefore
Fu-Xian Yi et al.
Biology of reproduction, 82(1), 66-75 (2009-09-11)
Endothelium-mediated vasodilation is specifically enhanced in uterine circulation during pregnancy, and production of nitric oxide (NO) is increased in response to a wide array of agonists. Uterine artery endothelial cells from nonpregnant (NP-UAECs) or pregnant (P-UAECs) ewes maintained in culture
Dongdong Yuan et al.
BMC anesthesiology, 21(1), 267-267 (2021-11-05)
Opioids have been identified by the World Health Organization to be 'indispensable for the relief of pain and suffering'. Side-effects, such as nausea, vomiting, postoperative delirium, and effects on breathing, of opioids have been well investigated; however, the influence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service