Skip to Content
Merck
All Photos(3)

Key Documents

SAB4500423

Sigma-Aldrich

Anti-ADORA2A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ADORA2, ADORA2A, Adenosine receptor A2a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 44 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADORA2A(135)

General description

Anti-ADORA2A Antibody detects endogenous levels of total ADORA2A protein.
Adenosine A2a receptor (ADORA2A) is a G protein-coupled receptor. It is highly expressed in the brain. It is located on human chromosome 22q11.23.

Immunogen

The antiserum was produced against synthesized peptide derived from human ADORA2A.

Immunogen Range: 321-370

Biochem/physiol Actions

Activation of the adenosine A2a receptor (ADORA2A) prevents T-cell proliferation and production of pro-inflammatory cytokines. It functions in increasing cytosolic cAMP levels. The protein has been associated with stimulation of liver ischemic preconditioning. ADORA2A is also linked to dermal fibrosis and scleroderma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacological Preconditioning by Adenosine A2a Receptor Stimulation: Features of the Protected Liver Cell Phenotype
Alchera E, et al.
BioMed Research International (2015)
Adenosine A2A receptor (A2AR) is a fine-tune regulator of the collagen1:collagen3 balance
Perez-Aso M, et al.
Purinergic Signaling, 9(4), 573-583 (2013)
A genetic variation in the ADORA2A gene modifies age at onset in Huntington's disease
Dhaenens CM, et al.
Neurobiology of Disease, 35(3), 474-476 (2009)
[Adenosine A2A receptor as a drug target for treatment of sepsis]
Sivak KV, et al.
Molekuliarnaia Biologiia, 50(2), 231-245 (2016)
Adenosine A(2A) receptor gene (ADORA2A) variants may increase autistic symptoms and anxiety in autism spectrum disorder.
Freitag CM
European Child & Adolescent Psychiatry, 19(1), 67-74 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service