Skip to Content
Merck
All Photos(3)

Documents

SAB4200750

Sigma-Aldrich

Anti-Gelsolin antibody, Mouse Monoclonal

clone GS-2C4, purified from hybridoma cell culture

Synonym(s):

Anit-AGEL, Anit-Actin-depolymerizing factor (ADF), Anit-GSN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

GS-2C4, monoclonal

form

buffered aqueous solution

mol wt

~85 kDa

species reactivity

rabbit, canine, human, mouse, monkey

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 1-2.5 μg/mL using MDCK cell line extract
immunofluorescence: suitable
immunohistochemistry: 10-20 μg/mL using heat-retrieved formalin-fixed, paraffin-embedded human pancreas sections
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSN(2934)

General description

Anti-Gelsolin Antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the GS-2C4 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Gelsolin operates in at least two variant forms encoded by a single gene and is located both intracellular (in cytosol and mitochondria) and extracellular (in blood plasma).

Immunogen

Human plasma gelsolin

Application

Anti-Gelsolin antibody, Mouse Monoclonal has been used in:
  • immunoblotting
  • immunofluorescence
  • immunohistochemistry
  • immunoprecipitation

Biochem/physiol Actions

Gelsolin, also known as Actin-depolymerizing factor (ADF), GSN, AGEL or Brevin, is a calcium-dependent actin-binding protein. Gelsolin acts as a main modulator of actin filament formation and disassembly and is involved in a variety of cellular mechanisms including apoptosis, signal transduction, transcriptional regulation and epigenetic processes. Plasma gelsolin differs in its sequence from the cytoplasmic variant in possessing a 25-amino acid N-terminal extension. The main function of the cytoplasmic gelsolin is to remodel the actin cytoskeleton. Plasma gelsolin′s main role is to scan for actin fibrils that are released by injured tissue into the bloodstream and to bind them in a scavenger mode preventing actin from increasing the blood viscosity. Gelsolin levels correlate with inflammation and several diseases and pathologies, including cancer, infection, cardiac injury, pulmonary diseases, Alzheimer′s disease and aging thus making it a potential biomarker of diagnostic and prognostic values.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteomic differences between hepatocellular carcinoma and nontumorous liver tissue investigated by a combined gel-based and label-free quantitative proteomics study
Megger DA, et al.
Molecular and Cellular Proteomics, 12(7), 2006-2020 (2013)
Topological assignment of the N-terminal extension of plasma gelsolin to the gelsolin surface
Ulrike F, et al.
The Biochemical Journal, 385(3), 659-665 (2005)
Membrane cholesterol, lateral mobility, and the phosphatidylinositol 4, 5-bisphosphate-dependent organization of cell actin
Kwik J, et al.
Proceedings of the National Academy of Sciences of the USA, 100(24), 13964-13969 (2003)
Gelsolin amyloidosis: genetics, biochemistry, pathology and possible strategies for therapeutic intervention
Solomon JP, et al.
Critical Reviews in Biochemistry and Molecular Biology, 47(3), 282-296 (2012)
Vesicular egress of non-enveloped lytic parvoviruses depends on gelsolin functioning
Bar S, et al.
PLoS Pathogens, 4(8), e1000126-e1000126 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service