Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500002

Sigma-Aldrich

Anti-TTBK1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BDTK, Anti-Brain derived tau kinase, Anti-Tau tubulin kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 145-151 kDa

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TTBK1(84630)

General description

Tau tubulin kinase 1 (TTBK1) is a serine/threonine and tyrosine kinase. It belongs to the casein kinase 1 superfamily and is neuron-specific. The encoded protein has a kinase domain, a unique polyglutamate domain and several PEST (proline–glutamate–serine–threonine) sequences. The gene encoding TTBK1 is localized on human chromosome 6p21.1.

Immunogen

TTBK1 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human TTBK1.

Application

Anti-TTBK1 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Tau tubulin kinase 1 (TTBK1) is responsible for phosphorylation of TDP-43 (TAR DNA binding protein), a protein associated with amyotrophic lateral sclerosis and frontotemporal lobar dementia. High expression or abundance of TTBK1 might be associated with neurodegeneration in these diseases. It also phosphorylates tau protein and might be associated with Alzheimer′s disease progression.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Tau tubulin kinase (TTBK1) belongs to the casein kinase 1 superfamily and is involved in the phosphorylation of specific serine/threonine residues in paired helical filaments of the tau protein. It is specifically expressed in the brain and induces tau aggregation in human neuronal cells in a dose-dependent manner. TTBK1 levels have been found to be upregulated in the brains of Alzheimer′s disease (AD) patients, and mice expressing human TTBK1 protein showed significant age-dependent memory impairment. These mice displayed increased levels of the CDK5 activators p25 and p35, and reduced levels of the NMDA receptor types 2B and 2D, suggesting that TTBK1 may play a role in memory dysfunction in AD patients. Multiple isoforms of TTBK1 are known to exist. This antibody is predicted to not cross-react with TTBK2.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500002.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The structure of human tau-tubulin kinase 1 both in the apo form and in complex with an inhibitor.
Kiefer SE
Acta Crystallographica. Section F, Structural Biology Communications (2014)
The Tau Tubulin Kinases TTBK1/2 Promote Accumulation of Pathological TDP-43
Nicole F
PLoS Genetics (2014)
The tau tubulin kinases TTBK1/2 promote accumulation of pathological TDP-43.
Liachko NF
PLoS Genetics (2014)
Nicole F Liachko et al.
PLoS genetics, 10(12), e1004803-e1004803 (2014-12-05)
Pathological aggregates of phosphorylated TDP-43 characterize amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD-TDP), two devastating groups of neurodegenerative disease. Kinase hyperactivity may be a consistent feature of ALS and FTLD-TDP, as phosphorylated TDP-43 is not observed in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service