Skip to Content
Merck
All Photos(1)

Key Documents

S6135

Sigma-Aldrich

Somatostatin 28

≥97% (HPLC)

Synonym(s):

Prosomatostatin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C137H207N41O39S3
CAS Number:
Molecular Weight:
3148.56
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

powder

mol wt

3148.56

storage condition

(Keep container tightly closed in a dry and well-ventilated place)

concentration

>70% (peptide content)

technique(s)

activity assay: suitable

solubility

water: 1 mg/mL, clear, colorless

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(C)NC(=O)C1CCCN1C(=O)C(CC(N)=O)NC(=O)C(CO)NC(=O)C(CC(N)=O)NC(=O)C(C)NC(=O)C(N)CO)C(=O)NC(C)C(=O)N2CCCC2C(=O)NC(CCCNC(N)=N)C(=O)NC(CCC(O)=O)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCCCN)C(=O)NC(C)C(=O)NCC(=O)NC3CSSCC(NC(=O)C(CO)NC(=O)C(NC(=O)C(Cc4ccccc4)NC(=O)C(NC(=O)C(CCCCN)NC(=O)C(Cc5c[nH]c6ccccc56)NC(=O)C(Cc7ccccc7)NC(=O)C(Cc8ccccc8)NC(=O)C(CC(N)=O)NC(=O)C(CCCCN)NC3=O)C(C)O)C(C)O)C(O)=O

InChI

1S/C137H207N41O39S3/c1-69(154-113(194)82(37-19-22-47-138)159-115(196)85(40-25-50-149-136(145)146)160-118(199)87(44-45-106(188)189)163-116(197)86(41-26-51-150-137(147)148)164-130(211)101-43-27-52-177(101)133(214)72(4)156-114(195)88(46-54-218-7)158-110(191)71(3)155-129(210)100-42-28-53-178(100)134(215)95(61-104(144)186)171-126(207)96(65-180)172-124(205)93(59-102(142)184)165-111(192)70(2)153-112(193)80(141)64-179)109(190)152-63-105(187)157-98-67-219-220-68-99(135(216)217)174-127(208)97(66-181)173-132(213)108(74(6)183)176-125(206)91(57-77-33-15-10-16-34-77)170-131(212)107(73(5)182)175-119(200)84(39-21-24-49-140)161-122(203)92(58-78-62-151-81-36-18-17-35-79(78)81)168-121(202)90(56-76-31-13-9-14-32-76)166-120(201)89(55-75-29-11-8-12-30-75)167-123(204)94(60-103(143)185)169-117(198)83(162-128(98)209)38-20-23-48-139/h8-18,29-36,62,69-74,80,82-101,107-108,151,179-183H,19-28,37-61,63-68,138-141H2,1-7H3,(H2,142,184)(H2,143,185)(H2,144,186)(H,152,190)(H,153,193)(H,154,194)(H,155,210)(H,156,195)(H,157,187)(H,158,191)(H,159,196)(H,160,199)(H,161,203)(H,162,209)(H,163,197)(H,164,211)(H,165,192)(H,166,201)(H,167,204)(H,168,202)(H,169,198)(H,170,212)(H,171,207)(H,172,205)(H,173,213)(H,174,208)(H,175,200)(H,176,206)(H,188,189)(H,216,217)(H4,145,146,149)(H4,147,148,150)

InChI key

GGYTXJNZMFRSLX-UHFFFAOYSA-N

Gene Information

human ... SST(6750)

Amino Acid Sequence

Ser-Ala-Asn-Ser-Asn-Pro-Ala-Met-Ala-Pro-Arg-Glu-Arg-Lys-Ala-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys [Disulfide Bridge: 17-28]

General description

Research area: Neuroscience

The SST gene is mapped to human chromosome 3q27.3. Both SST-14 and SST-28 are cyclic peptide.
The somatostatin gene codes for a peptide hormone containing 14 and 28 amino acid moieties. SST-14 is highly expressed in the central nervous system and peripheral organs. SST-28 is mainly expressed in enteroendocrine cells of the intestine.

Application

Somatostatin 28 has been used:

  • to study the effects of somatostatin in regulating hepatic growth hormone sensitivity
  • to study its effects on insulin-like growth factor-I receptor expression in the gills of Oncorhynchus mykiss (a teleost fish)
  • to assess the effects of somatostatinon GH (growth hormone)-stimulated IGF-I (insulin-like growth factor-I) expression

Biochem/physiol Actions

Somatostatin action is delivered with help of five different receptor subtypes (sst1 to sst5).
Somatostatin plays an important role in regulating the metabolism of other hormones such as growth hormone, insulin, IGF-1 (insulin-like growth factor 1), glucagon, glucagon-like peptide and gastrin. Somatostatin has both regulatory and inhibitory functions. It retards gastrointestinal motility and gallbladder contraction, suppresses gut exocrine secretion and reduces epithelial proliferation. Somatostatin is associated with intestinal nutrient absorption and blood flow regulation. This peptide hormone is believed to inhibit cell proliferation and promote apoptosis. Somatostatin acts as a neurotransmitter and neuromodulator, and is also associated in maintaining water balance.Pituitary somatostatin receptor signaling is implicated in the inhibition of adenylyl cyclase- cyclic adenosine 3′,5′-monophosphate (cAMP)- protein kinase A (PKA) pathway. Pituitary somatostatin activity is also involved in regulation of various pathways including guanylyl cyclase and nitric oxide (NO), mitogen-activated protein kinase (MAPK) and protein kinase B (PI3K/Akt).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Modena et al.
Cancers, 15(13) (2023-07-14)
Somatostatin receptor (SSTR) agonists have been extensively used for treating neuroendocrine tumors. Synthetic therapeutic agonists showing selectivity for SSTR2 (Octreotide) or for SSTR2 and SSTR5 (Pasireotide) have been approved for the treatment of patients with acromegaly and Cushing's syndrome, as
Somatostatin regulates hepatic growth hormone sensitivity by internalizing growth hormone receptors and by decreasing transcription of growth hormone receptor mRNAs
Nicole M
Physiology (Bethesda, MD) (2007)
V Martínez et al.
The Journal of pharmacology and experimental therapeutics, 293(3), 1099-1105 (2000-06-28)
We previously showed that the somatostatin receptor 5 (sst(5))-preferring agonist BIM-23052 injected intracisternally (i.c. ; 0.8 nmol/rat) stimulated gastric emptying of a non-nutrient meal in conscious rats. In this study, we investigated the neural pathways and specificity of BIM-23052 action.
Somatostatin regulates hepatic growth hormone sensitivity by internalizing growth hormone receptors (GHR) and by decreasing transcription of GHR mRNAs
Very NM and Sheridan MA
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology (2007)
Anat Ben-Shlomo et al.
Trends in endocrinology and metabolism: TEM, 21(3), 123-133 (2010-02-13)
Somatotropin-release inhibitory factor (SRIF) is a major regulator of pituitary function, mostly inhibiting hormone secretion and to a lesser extent pituitary cell growth. Five SRIF receptor subtypes (SSTR1-5) are ubiquitously expressed G-protein coupled receptors. In the pituitary, SSTR1, 2, 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service