Skip to Content
Merck
All Photos(5)

Key Documents

RDD004

Sigma-Aldrich

PIPES

anhydrous, free-flowing, Redi-Dri, ≥99%

Synonym(s):

1,4-Piperazinediethanesulfonic acid, Piperazine-1,4-bis(2-ethanesulfonic acid), Piperazine-N,N′-bis(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O6S2
CAS Number:
Molecular Weight:
302.37
Beilstein:
817713
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

anhydrous

product line

Redi-Dri

Assay

≥99%

form

crystalline powder

quality

free-flowing

loss

≤1.0% loss on drying, 110 degrees Celcius

useful pH range

6.1-7.5

pKa (25 °C)

6.8

mp

>300 °C (lit.)

solubility

1 M NaOH: 20 + 80 mL g, clear, colorless

SMILES string

OS(=O)(=O)CCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O6S2/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16/h1-8H2,(H,11,12,13)(H,14,15,16)

InChI key

IHPYMWDTONKSCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leticia A Montoya et al.
Analytical chemistry, 86(12), 6032-6039 (2014-05-24)
Sulfhydryl-containing compounds, including thiols and hydrogen sulfide (H2S), play important but differential roles in biological structure and function. One major challenge in separating the biological roles of thiols and H2S is developing tools to effectively separate the reactivity of these
Veronique Adoue et al.
Molecular systems biology, 10, 754-754 (2014-10-19)
Most complex disease-associated genetic variants are located in non-coding regions and are therefore thought to be regulatory in nature. Association mapping of differential allelic expression (AE) is a powerful method to identify SNPs with direct cis-regulatory impact (cis-rSNPs). We used
Kalina Makowiecki et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(32), 10780-10792 (2014-08-08)
Repetitive transcranial magnetic stimulation (rTMS) is increasingly used as a treatment for neurological and psychiatric disorders. Although the induced field is focused on a target region during rTMS, adjacent areas also receive stimulation at a lower intensity and the contribution
Nicholas Light et al.
Epigenetics, 9(9), 1238-1251 (2014-07-24)
Allele-specific (AS) assessment of chromatin has the potential to elucidate specific cis-regulatory mechanisms, which are predicted to underlie the majority of the known genetic associations to complex disease. However, development of chromatin landscapes at allelic resolution has been challenging since
Matthew D Hammers et al.
Analytical chemistry, 86(14), 7135-7140 (2014-06-18)
Hydrogen sulfide (H2S) is an integral signaling molecule in biology with complex generation, translocation, and metabolism processes that are intertwined with cellular thiols. Differentiating the complex interplay between H2S and biological thiols, however, remains challenging due to the difficulty of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service