Skip to Content
Merck
All Photos(2)

Documents

RAB0290

Sigma-Aldrich

Human IL-22 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8 pg/mL
standard curve range: 6.55-1600 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL22(50616)

General description

Human Interleukin-22 (IL-22) enzyme-linked immunosorbent assay (ELISA) Kit is an in vitro assay for the quantitative measurement of human IL-22 in biological samples, such as serum, plasma, cell culture supernatants, urine, and/or cell and tissue lysates. IL-22 belongs to the family of IL-10 of cytokines. It is produced by the γδ T cells, CD4+ T helper cells, natural killer T cells, and innate lymphoid cells. It is involved in the activation of signal transducer and activator of transcription 3 and 1 (STAT3/1). IL-22 possesses stage-specific, pro-and anti-tumor effects.

Immunogen

Recombinant Human IL-22

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chrissie Lim et al.
Science signaling, 9(447), ra95-ra95 (2016-09-30)
The cytokine interleukin-22 (IL-22), which is a member of the IL-10 family, is produced exclusively by immune cells and activates signal transducer and activator of transcription 3 (STAT3) in nonimmune cells, such as hepatocytes, keratinocytes, and colonic epithelial cells, to
P Cordiali-Fei et al.
Mediators of inflammation, 2014, 236060-236060 (2014-08-20)
The therapeutic management of psoriasis includes conventional treatments as well as the new generation of highly effective TNF-α inhibitors. However, psoriasis has proven to be a complex therapeutic challenge and treatment failures are not uncommon. Thus, laboratory biomarkers of disease
Rosaria Maddalena Ruggeri et al.
Hormones (Athens, Greece), 13(3), 338-344 (2014-08-01)
Hashimoto's thyroiditis (HT) is considered to be a Th1-related autoimmune disease (AID). Recent studies revealed that Th17 lymphocytes (producing mostly IL-17, IL-21 and IL-22) play a major role in numerous AIDs commonly thought to be Th1 diseases. More recently, another
Daniel E Dulek et al.
Infection and immunity, 82(9), 3723-3739 (2014-06-25)
The Th17 cytokines interleukin-17A (IL-17A), IL-17F, and IL-22 are critical for the lung immune response to a variety of bacterial pathogens, including Klebsiella pneumoniae. Th2 cytokine expression in the airways is a characteristic feature of asthma and allergic airway inflammation.
Mohan S Maddur et al.
Nature communications, 5, 4092-4092 (2014-06-10)
Dendritic cells (DCs) play a critical role in immune homeostasis by regulating the functions of various immune cells, including T and B cells. Notably, DCs also undergo education on reciprocal signalling by these immune cells and environmental factors. Various reports

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service