Skip to Content
Merck
All Photos(1)

Key Documents

P6998

Sigma-Aldrich

Protein Kinase A Catalytic Subunit β, Active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PKA Catalytic Subunit β, Active human, PKA-Cβ), cAMP-dependent protein kinase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

Assay

≥80% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

≥10 nmol/min·mg

mol wt

protein apparent mol wt ~65 kDa (including N-terminal GST-tag of ~26kDa)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKACB(5567)

General description

Protein kinase cAMP-activated catalytic subunit β (PRKACB) is one of the catalytic subunits of the serine/threonine kinase, protein kinase A. The gene encoding it is localized on human chromosome 1p36.1.

The molecular weight of about 65kDa is confirmed by the SDS-PAGE gel. The product was generated by expressing recombinant full-length human PKAcβ2 with an additional N-terminal GST tag by baculovirus in Sf9 insect cells. The gene accession number is NM_002731. The resulting protein is 351aa cAMP-dependent protein kinase catalytic subunit beta isoform 2 (NP_002722), or according to UniProt, Isoform 1 (P22694-1).

Application

Kinase activity is measured as the molar amount of phosphate incorporated into the CREBtide substrate peptide per minute per mg protein at 30 °C using a final concentration of 50 μM [32P] ATP.

Biochem/physiol Actions

cAMP-dependent protein kinase (PKA; EC 2.7.1.37) is an essential enzyme in the signaling pathway of the second messenger cAMP. Through phosphorylation of target proteins, PKA controls many biochemical events in the cell including regulation of metabolism, ion transport, and gene transcription.

Unit Definition

One unit will incorporate one nanomole of phosphate into the Crebtide (KRREILSRRPSYR) substrate per minute at 30 deg C and pH 7.2 using a final concentration of 50 μM [32P] ATP.

Physical form

Supplied as a solution of 5 μg in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF and 25% glycerol.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association between PKA gene polymorphism and NTDs in high risk Chinese population in Shanxi.
Wu J, et.al
International Journal of Clinical and Experimental Pathology, 6(12), 2968-2974 (2013)
Jian Wu et al.
International journal of clinical and experimental pathology, 6(12), 2968-2974 (2013-12-03)
This study aimed to investigate the single nucleotide polymorphisms (SNPs) of PKA and neural tube defects (NTDs) in Chinese population. A total of 183 NTDs cases and 200 healthy controls were used in this study. 7 selected single nucleotide polymorphism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service