Skip to Content
Merck
All Photos(1)

Key Documents

P6624

Sigma-Aldrich

PS-1145 dihydrochloride

≥98% (HPLC), solid

Synonym(s):

N-(6-Chloro-9H-pyrido[3,4-b]indol-8-yl)-3-pyridinecarboxamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11ClN4O · 2HCl
CAS Number:
Molecular Weight:
395.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

yellow

solubility

DMSO: >3 mg/mL at ~60 °C, clear (with warming for 2 minutes)
methanol: water (1:1): ≥5 mg/mL at 60 °C (With warming for 5 minutes.)

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Clc1cc(NC(=O)c2cccnc2)c3[nH]c4cnccc4c3c1

InChI

1S/C17H11ClN4O.2ClH/c18-11-6-13-12-3-5-20-9-15(12)21-16(13)14(7-11)22-17(23)10-2-1-4-19-8-10;;/h1-9,21H,(H,22,23);2*1H

InChI key

QTDCBADLGJZBHP-UHFFFAOYSA-N

Application

PS-1145 dihydrochloride has been used as an IκB kinase (IKK) inhibitor:
  • in the culture medium to treat human epidermal keratinocytes (HEKs) and squamous carcinoma cells (SCCs) to study the role of IKK in Corynebacterium tuberculostearicum (C. t.)-elicited transcription of inflammatory mediators
  • to investigate the involvement of transcription factor p65 (RelA)/nuclear factor kappa-B (NF-κB) in the cluster of differentiation 28 (CD28)-mediated interleukin-17A (IL-17A) gene expression
  • to examine the involvement of NF-κB signaling in the cytotoxic effect of niclosamide on colorectal cancer (CRC) cells

Biochem/physiol Actions

PS-1145 is an IKB kinase (IKK) inhibitor.
PS-1145, a β-carboline derivative, blocks the phosphorylation and degradation of the inhibitor of nuclear factor kappa-β (NF-κB;). It also inhibits the subsequent activation of nuclear factor kappa-β (NF-κB) and thereby prevents the release of tumor necrosis factor-α (TNF-α) in lipopolysaccharide treated mice. PS-1145 hinders pro-inflammatory cytokine production and cell proliferation. It plays a potential therapeutic role in carcinogenesis in multiple myeloma. PS-1145 exhibits anti-tumor activity on nasopharyngeal carcinoma (NPC) cell lines.

Features and Benefits

This compound is featured on the MAPKKKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Yemelyanov et al.
Oncogene, 25(3), 387-398 (2005-09-20)
A key antiapoptotic transcription factor, nuclear factor kappa-B (NF-kappaB), is known to be critically important for tumor cell growth, angiogenesis and development of metastatic lesions. We and others showed previously that NF-kappaB transcription factor was constitutively activated in androgen-independent prostate
Lloyd T Lam et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 11(1), 28-40 (2005-01-27)
Constitutive activation of the NF-kappaB pathway is required for survival of the activated B cell-like (ABC) subgroup of diffuse large B-cell lymphoma (DLBCL). Here we show that a small molecule IkappaB kinase (IKK) inhibitor, PS-1145, and related compounds are toxic
Takuya Osada et al.
Cancer research, 71(12), 4172-4182 (2011-05-03)
Wnt/β-catenin pathway activation caused by adenomatous polyposis coli (APC) mutations occurs in approximately 80% of sporadic colorectal cancers (CRC). The antihelminth compound niclosamide downregulates components of the Wnt pathway, specifically Dishevelled-2 (Dvl2) expression, resulting in diminished downstream β-catenin signaling. In
Sameek Roychowdhury et al.
Journal of the National Cancer Institute, 96(19), 1447-1457 (2004-10-07)
Immune-compromised individuals are at increased risk for developing aggressive Epstein-Barr virus (EBV)-associated lymphoproliferative disorders after primary EBV infection or for reactivation of a preexisting latent EBV infection. We evaluated the effect of depsipeptide, a histone deacetylase inhibitor, on EBV-positive lymphoblastoid
Ian C Henrich et al.
Cancer research, 81(8), 2171-2183 (2021-02-10)
Ewing sarcoma is the second most common pediatric bone cancer, with a 5-year survival rate for metastatic disease of only 20%. Recent work indicates that survival is strongly correlated with high levels of tumor-infiltrating lymphocytes (TIL), whose abundance is associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service