Skip to Content
Merck
All Photos(4)

Documents

O4139

Sigma-Aldrich

Orlistat

≥98% (HPLC), solid, lipase inhibitor

Synonym(s):

(−)-Tetrahydrolipstatin, N-Formyl-L-leucine (1S)-1-[[(2S,3S)-3-hexyl-4-oxo-2-oxetanyl]methyl]dodecyl ester, Ro-18-0647

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H53NO5
CAS Number:
Molecular Weight:
495.73
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

product name

Orlistat, ≥98%, solid

biological source

synthetic (organic)

Assay

≥98%

form

solid

color

white

mp

<50 °C

solubility

DMSO: 19 mg/mL

originator

Roche

storage temp.

2-8°C

SMILES string

CCCCCCCCCCC[C@@H](C[C@@H]1OC(=O)[C@H]1CCCCCC)OC(=O)[C@H](CC(C)C)NC=O

InChI

1S/C29H53NO5/c1-5-7-9-11-12-13-14-15-16-18-24(34-29(33)26(30-22-31)20-23(3)4)21-27-25(28(32)35-27)19-17-10-8-6-2/h22-27H,5-21H2,1-4H3,(H,30,31)/t24-,25-,26-,27-/m0/s1

InChI key

AHLBNYSZXLDEJQ-FWEHEUNISA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Orlistat acts as a gastric lipase inhibitor that reduces fat absorption.

Application

Orlistat has been used as an inhibitor:
  • of fatty acid synthase in long-term estrogen-deprived (LTED) variant cell lines
  • of lipase in screening lipase inhibition by raspberry extract
  • in in vitro pancreatic lipase activity assay

Biochem/physiol Actions

Orlistat impairs intestinal fat absorption and is effective in weight management. It is regarded as a safe drug for treating obesity. It delays the progression of type 2 diabetes mellitus especially in obese and improves glycemic parameters. Long term usage of orlistat results in improved weight reduction and it may not contribute in colorectal carcinogenesis.
Orlistat, used in obesity research, is a pancreatic lipase inhibitor that acts locally in the gastrointestinal tract to inhibit lipase.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Other Notes

Solution is not completely clear, small particles may remain floating.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibitory effect of raspberries on starch digestive enzyme and their antioxidant properties and phenolic composition
Zhang L, et al.
Food Chemistry, 119(2), 592-599 (2010)
Effect of orlistat on weight and body composition in obese adolescents: a randomized controlled trial
Chanoine JP, et al.
JAMA : The Journal of the American Medical Association, 293(23), 2873-2883 (2005)
Shiva Kant et al.
Biochimica et biophysica acta, 1840(1), 294-302 (2013-09-26)
Orlistat, a fatty acid synthase (FASN) inhibitor, has been demonstrated to inhibit tumor cell survival. However, the mechanism(s) of its tumor growth retarding action against malignancies of hematological origin remains unclear. It is also not understood if the antitumor action
Risk of colorectal cancer after initiation of orlistat: matched cohort study
Hong JL, et al.
BMJ (Clinical Research ed.), 347(12), f5039-f5039 (2013)
The effect of consumption of citrus fruit and olive leaf extract on lipid metabolism
Merola N, et al.
Nutrients, 9(10), 1062-1062 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service