Skip to Content
Merck
All Photos(1)

Documents

N3520

Sigma-Aldrich

Nutrient Mixture F-12 Ham

Kaighn′s Modification, with L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

Ham’s F-12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

form

powder

technique(s)

cell culture | mammalian: suitable

components

NaHCO3: no
sodium pyruvate: 0.22 g/L
HEPES: no
L-glutamine: yes
phenol red: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The nutrient mixture Ham′s F-12 is developed by Ham and it has higher levels of amino acids, vitamins and trace elements. Putrescine and linoleic acid are added to the formulation. Nutrient medium was originally designed for the serum-free growth of Chinese hamster ovary and lung cells. When used with whole or dialyzed serum or in combination with hormones and transferrin, it is widely used to grow mammalian and hybridoma cells.

Application

Nutrient Mixture F-12 Ham is used for the following applications:
  • HeLa (human cervix adenocarcinoma cells) were grown in this medium (nutrient mixture F-12 Ham) supplemented with 10% heat-inactivated fetal calf serum
  • Used in cell culture for different cells during maintenance and incubation (i.e. A549 cell line was maintained as a monolayer in nutrient mixture F-12 HAM Medium along with other components)
  • Used in media for the propagation of Cowdria ruminantium
  • Used during preparation of primary mixed glial cultures

Kaighn′s modification of Ham′s F-12 and Coon′s F-12 has increased concentrations of amino acids and pyruvate, as well as modified salts (Konigsbergs). This medium was designed to support the growth of differentiated rat and chicken cells, and primary human liver cells.

Quantity

Formulated to contain 11.1 grams of powder per liter of medium.

Reconstitution

Supplement with 2.5 g/L sodium bicarbonate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Yang et al.
Environmental science and pollution research international, 25(1), 867-876 (2017-10-27)
This research is designed to investigate the regulatory effect of miR-18a to the target gene connective tissue growth factor (CTGF, or CCN2), by participating in TGF-β1 signaling pathway and explore the pathogenic mechanism of miR-18a in pulmonary injury induced by
A Tansu Koparal et al.
Cytotechnology, 43(1-3), 149-154 (2008-11-13)
Carvacrol, the predominant monoterpene in many essential oils of Labitae including Origanum, Satureja, Thymbra, Thymus, and Corydothymus has substantial antibacterial, antifungal, antihelmintic, insecticial, analgesic and antioxidant activities. Approximately 75% of lung cancer is non-small cell carcinoma (NSCLC) which comprises several
Anna Mero et al.
Journal of controlled release : official journal of the Controlled Release Society, 125(2), 87-95 (2007-11-23)
Poly(2-ethyl-2-oxazoline) (POZ) was synthesized by living cationic ring-opening polymerization under microwave irradiation yielding polymers with low polydispersity indices (PDI, 1.15). The polymerization was quenched with sodium carbonate yielding a hydroxyl end-group with a high degree of functionality. The hydroxyl group
Haiping Wang et al.
Molecular medicine reports, 15(2), 743-749 (2016-12-14)
B23, a multifunctional nucleolar protein, is overexpressed in numerous cancers and is associated with tumorigenesis. However, the clinical significance and potential role of B23 in bladder urothelial carcinoma remains to be elucidated. The present study observed that the mRNA and
Hanli Guo et al.
Journal of advanced research, 28, 255-267 (2020-12-29)
Introduction: Disruptions of extracellular matrix (ECM) degradation homeostasis play a significant role in the pathogenesis of osteoarthritis (OA). Matrix metalloproteinase 13 (MMP13) and collagen Ⅱ are important components of ECM. Earlier we found that quercitrin could significantly decrease MMP13 gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service