Skip to Content
Merck
All Photos(2)

Key Documents

M2786

Sigma-Aldrich

3-Maleimidobenzoic acid N-hydroxysuccinimide ester

crystalline

Synonym(s):

MBS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10N2O6
CAS Number:
Molecular Weight:
314.25
Beilstein:
1505254
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

form

crystalline

Quality Level

reaction suitability

reagent type: cross-linking reagent

mp

175-177 °C (lit.)

solubility

DMSO: ≤20 mg/mL

functional group

NHS ester
maleimide

storage temp.

−20°C

SMILES string

O=C1CCC(N1OC(C2=CC=CC(N3C(C=CC3=O)=O)=C2)=O)=O

InChI

1S/C15H10N2O6/c18-11-4-5-12(19)16(11)10-3-1-2-9(8-10)15(22)23-17-13(20)6-7-14(17)21/h1-5,8H,6-7H2

InChI key

LLXVXPPXELIDGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

may require addition of solvent to coupling buffer to at least 5% to maintain solubility

Application

3-Maleimidobenzoic acid N-hydroxysuccinimide ester (MBS) has been used as a protein cross-linker. It has been used for the stabilization of actin filaments in plant tissue samples.
3-Maleimidobenzoic acid N-hydroxysuccinimide ester has been used:
  • in the fixation of pollen in Arabidopis
  • as a component of Murashige and Skoog (MS) medium to stabilize F-actin in Eucalyptus root hairs
  • in the fixation of F-actin labelled pollen tubes of Lilium davidii

Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.0, followed by coupling to compounds containing sulfhydryl by thioether at same pH. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.

Biochem/physiol Actions

3-Maleimidobenzoic acid N-hydroxysuccinimide is used as a fixative for the preservation of pollen tube F-actin distribution. It imposes less damage when used on pollen tubes.

Other Notes

The aryl maleimide is less stable than alkyl maleimide linkers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Visualization of Actin Organization and Quantification in Fixed Arabidopsis Pollen Grains and Tubes
Qu X, et al.
Bio-protocol, 10(01), 204-211 (2020)
Andrii Slonchak et al.
BMC genomics, 20(1), 474-474 (2019-06-12)
Extracellular vesicles (EVs) are small membrane vesicles secreted by the cells that mediate intercellular transfer of molecules and contribute to transduction of various signals. Viral infection and action of pro-inflammatory cytokines has been shown to alter molecular composition of EV
Ilenia Papa et al.
Nature, 547(7663), 318-323 (2017-07-13)
Protective high-affinity antibody responses depend on competitive selection of B cells carrying somatically mutated B-cell receptors by follicular helper T (T
Jeong Hwan Seong et al.
Genes, 9(7) (2018-07-04)
The 14-3-3 family of proteins performs key regulatory functions in phosphorylation-dependent signaling pathways including cell survival and proliferation, apoptosis, regulation of chromatin structure and autophagy. In this study, the zeta isoform of 14-3-3 proteins (designated as Tm14-3-3ζ) was identified from
Comparison of F-actin fluorescent labeling methods in pollen tubes of Lilium davidii
Wang L, et al.
Plant Cell Reports, 24(5), 266-270 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service