Skip to Content
Merck
All Photos(1)

Documents

M0199

Sigma-Aldrich

Mdivi-1

≥98% (HPLC), powder, mitochondrial division DRP inhibitor

Synonym(s):

3-(2,4-Dichloro-5-methoxyphenyl)-2,3-dihydro-2-thioxo-4(1H)-quinazolinone, 3-(2,4-Dichloro-5-methoxyphenyl)-2-sulfanyl-4(3H)-quinazolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10Cl2N2O2S
CAS Number:
Molecular Weight:
353.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Mdivi-1, ≥98% (HPLC), powder

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >20 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

COc1cc(N2C(S)=Nc3ccccc3C2=O)c(Cl)cc1Cl

InChI

1S/C15H10Cl2N2O2S/c1-21-13-7-12(9(16)6-10(13)17)19-14(20)8-4-2-3-5-11(8)18-15(19)22/h2-7H,1H3,(H,18,22)

InChI key

NZJKEVWTYMOYOR-UHFFFAOYSA-N

General description

Mitochondrial Division Inhibitor 1 (Mdivi-1) is a quinazolinone derivative and is cell permeable. It traverses blood-brain barrier and elicits protective functionality in heart and brain ischemia-reperfusion injury.

Application

Mdivi-1 has been used:
  • in embryonic thoracic aorta A7r5 cells to inhibit cell migration and proliferation
  • in mitochondrial network reshaping and reactive oxygen species (ROS) production studies in oligodendrocyte precursor cells (OPCs)
  • to induce mitochondrial damage in lung fibroblasts

Biochem/physiol Actions

Mdivi-1 is a cell-permeable selective inhibitor of mitochondrial division DRP (dynamin-related GTPase) and inhibitor of the mitochondrial division dynamin (Dnm1). Mitochondrial fusion and division play important roles in the regulation of apoptosis. Mdivi-1 is the first selective inhibitor of mitochondrial division dynamins. In principle, Mdivi-1 represents a class of therapeutics for stroke, myocardial infarction, and neurodegenerative diseases.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

To mdivi-1 or not to mdivi-1: Is that the question?
Smith G and Gallo G
Developmental neurobiology, 77(11), 1260-1268 (2017)
Impaired mitophagy leads to cigarette smoke stress-induced cellular senescence: implications for chronic obstructive pulmonary disease
Ahmad T, et al.
Faseb Journal, 29(7), 2912-2929 (2015)
Ze Liu et al.
Antioxidants & redox signaling, 30(15), 1797-1816 (2018-06-13)
Mitochondrial fragmentation is a crucial mechanism contributing to tubular cell apoptosis during acute kidney injury (AKI). However, the mechanism of modulating mitochondrial dynamics during AKI remains unclear. Numb is a multifunction adaptor protein that is expressed in renal tubules. The
Da Zhang et al.
PloS one, 13(5), e0197266-e0197266 (2018-05-17)
Hypoxia-mediated pancreatic beta cell death is one of the main causes of pancreatic beta celldeath, which leads to the loss of functional pancreatic beta cell mass and type 1 diabetes andtype 2 diabetes.However, the molecular mechanisms that control life and
The putative Drp1 inhibitor mdivi-1 is a reversible mitochondrial complex I inhibitor that modulates reactive oxygen species
Bordt EA, et al.
Developmental Cell, 40(6), 583-594 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service