Skip to Content
Merck
All Photos(3)

Documents

L8005

Sigma-Aldrich

β-Lactoglobulin B from bovine milk

≥90% (PAGE)

Synonym(s):

β-LG, Bos d 5, beta-LG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

bovine milk

Assay

≥90% (PAGE)

form

powder

mol wt

18,276 Da by calculation

technique(s)

HPLC: suitable
electrophoresis: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... LGB(280838)

General description

β-Lactoglobulin B (β-LG B), a isoform of β-Lactoglobulin, is a small protein of 162 amino acids with a molecular mass of 18.2 kDa and optimum pH of 5.2. It is present in high level in casein and is relatively low in raw bovine milk. β-LG has eight-stranded β-barrel (strands A-H) succeeded by a three-turn α-helix and a final β-strand (strand I), that forms part of the dimerization interface.
Milk from dairy cows contains the protein β-lactoglobulin (BLG). It naturally occurs in a number of genetic variants, and the most prevalent bovine variants are known as BLG A and BLG B.

Application

β-Lactoglobulin B from bovine milk has been used as:
  • a protein standard in SDS-polyacrylamide electrophoresis for quantification of milk protein fractions
  • in the calibration of reversed phase- high-performance liquid chromatography (HPLC) for caseins quantification
  • for immobilization on the biosensor surface and a calibration standards in biosensor assay

β-Lactoglobulin was used in the identification of the genetic variants of κ-casein in milk by isoelectric focusing electrophoresis.

Biochem/physiol Actions

β-Lactoglobulin B (β-LG B) show less inhibitory effect on the Staphylaococcus sp compared to β-LG A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antimicrobial activity of bovine beta-lactoglobulin against mastitis-causing bacteria
Chaneton L, et al.
Journal of Dairy Science, 94(1), 138-145 (2011)
Huaying Zhao et al.
Current protocols in protein science, 101(1), e109-e109 (2020-07-03)
Sedimentation velocity analytical ultracentrifugation is a powerful classical method to study protein self-association processes in solution based on the size-dependent macromolecular migration in the centrifugal field. This technique can elucidate the assembly scheme, measure affinities ranging from picomolar to millimolar
The beta-lactoglobulin content of bovine milk: Development and application of a biosensor immunoassay
Indyk HE, et al.
International dairy journal, 73, 68-73 (2017)
Analysis of alpha-lactalbumin, beta-lactoglobulin A and B in whey protein powder, colostrum, raw milk, and infant formula by CE and LC
Ding X, et al.
Dairy science & technology, 91(2), 213-225 (2011)
Milk protein fractions strongly affect the patterns of coagulation, curd firming, and syneresis
Amalfitano N, et al.
Journal of Dairy Science, 102(4), 2903-2917 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service