Skip to Content
Merck
All Photos(1)

Documents

L2777

Sigma-Aldrich

Lisinopril

ACE Inhibitor

Synonym(s):

(S)-Nα-(1-Carboxy-3-phenylpropyl)-Lys-Pro

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204
NACRES:
NA.26

General description

Lisinopril is a nonsulfhydryl angiotensin-converting enzyme (ACE) inhibitor. It is used in the treatment of heart failure and hypertension. Lisinopril is an antihypertensive and anticongestive agent, like other members of its family. It is water-soluble and possesses weak chelating properties.

Application

Lisinopril has been used as an angiotensin-converting enzyme (ACE) inhibitor:
  • in combination with spironolactone, to study their effects on cardiac and skeletal muscles in the Duchenne muscular dystrophy (DMD) mice model
  • standard in in vitro ACE inhibitory assay
  • as a negative control in ACE enzymatic assay

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A A Patchett et al.
Nature, 288(5788), 280-283 (1980-11-20)
Much current attention focuses on the renin-angiotensin system in relation to mechanisms controlling blood pressure and renal function. Recent demonstrations (ref. 1, ref. 2 and refs therein) that angiotensin-converting enzyme inhibitors show promising clinical antihypertensive properties have been of particular
M L Ricketts et al.
Clinical science (London, England : 1979), 96(6), 669-675 (1999-05-21)
In the kidney and colon 11beta-hydroxysteroid dehydrogenase type 2 (11beta-HSD2) inactivates cortisol to cortisone, thereby protecting the non-selective mineralocorticoid receptor from cortisol. Deficiency of 11beta-HSD2 results in cortisol-mediated sodium retention and hypertension, suggesting that the physiological regulation of 11beta-HSD2 in
E J Wallis et al.
Clinical pharmacology and therapeutics, 64(4), 439-449 (1998-11-03)
The antihypertensive response to angiotensin-converting enzyme (ACE) inhibitors may be attenuated by a compensatory decrease in atrial natriuretic factor production. If so, inhibition of atrial natriuretic factor breakdown by neutral endopeptidase (NEP) may enhance the antihypertensive effects of ACE inhibition.
Comparative Effects of Low and High Doses of the
Angiotensin-Converting Enzyme Inhibitor, Lisinopril, on
Morbidity and Mortality in Chronic Heart Failure
Milton Packer
Clinical and Investigative Medicine. Medecine Clinique Et Experimentale (2017)
V B Sørensen et al.
Clinical science (London, England : 1979), 95(6), 709-717 (1998-12-01)
1. Our objective was to compare the effect of a long-acting calcium antagonist (nisoldipine) compared with an angiotensin-converting enzyme inhibitor (lisinopril) on the non-neurogenic regulation of the microvascular blood flow in hypertensive Type I diabetes patients with diabetic nephropathy.2. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service