Skip to Content
Merck
All Photos(1)

Key Documents

I7638

Sigma-Aldrich

Indoxyl β-D-glucuronide cyclohexylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15NO7 · C6H13N
CAS Number:
Molecular Weight:
408.45
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98%

form

powder

optical activity

[α]20/D -80.0±5.0

technique(s)

HPLC: suitable

storage temp.

−20°C

SMILES string

OC1C(OC(C(O)C1O)C(O)=O)Oc2c[nH]c3ccccc23

InChI

1S/C14H15NO7/c16-9-10(17)12(13(19)20)22-14(11(9)18)21-8-5-15-7-4-2-1-3-6(7)8/h1-5,9-12,14-18H,(H,19,20)

InChI key

KUYNOZVWCFXSNE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fujita et al.
Rinsho byori. The Japanese journal of clinical pathology, 44(9), 895-898 (1996-09-01)
To establish a simple identification procedure for Escherichia coli, we developed a disk containing indoxyl-beta-D-glucuronide, the chromogenic substrate of beta-D-glucuronidase. Of 188 isolates of Enterobacteriaceae, 101 (97%) of 104 strains of E. coli and two (67%) of three strains of
C H Tsai et al.
Journal of agricultural and food chemistry, 49(5), 2137-2141 (2001-05-23)
With the availability of an ultraweak chemiluminescence analyzer, it is possible to monitor the production of a specific oxygen-derived reactive species, such as hydroxyl radical ((*)OH), whenever a suitable chemiluminescent probe is obtainable. Reported herein is the development of a
T Niwa et al.
Nephron, 74(1), 72-78 (1996-01-01)
We identified and quantified indoxyl-beta-D-glucuronide in uremic serum and urine to determine the metabolism of indoles including indoxyl sulfate in uremic patients. Serum levels of indoxyl-beta-D-glucuronide were markedly increased in undialyzed uremic patients, in patients on hemodialysis, and in patients
S Agatsuma et al.
Clinical nephrology, 45(4), 250-256 (1996-04-01)
The content of indoxyl-beta-D-glucuronide, which has been found in patients' plasma as a new indicator of renal failure, logarithmically correlated with that of 3-indoxyl sulfate (indican) in the plasma of hemodialysis patients, showing another weak correlation with beta(2)-microglobulin content. The
G J Delisle et al.
Journal of clinical microbiology, 27(4), 778-779 (1989-04-01)
A new compound, indoxyl-beta-D-glucuronide, was assessed as a substrate for the rapid detection of Escherichia coli in urine. Incorporation of this compound into MacConkey agar allowed the direct differentiation of E. coli as deep blue colonies distinct from lactose and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service