Skip to Content
Merck
All Photos(1)

Documents

I5896

Sigma-Aldrich

Interleukin-7 human

≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IL-7

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≤0.500 ng/mL EC50

mol wt

25 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

solubility

water: soluble 0.005-0.010 mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL7(3574)

General description

The interleukin-7 human (IL-7) gene is mapped to human chromosome 8q21.13. It is a non-hematopoietic cell-derived cytokine. IL-7 is majorly produced by the fibroblastic stromal cells, dendritic cells, thymus epithelial cells and skin keratinocytes. Structurally, IL-7 takes up an up-up-down-down 4-helix bundle topology and has two crossover loops.

Biochem/physiol Actions

IL-7 helix A is crucial for its interaction with the interleukin-7 human receptor (IL-7Rα). It is reported to enhance immunity during viral infections and may be useful in cancer immunotherapy. Mutation in the IL-7 gene is implicated in CD4+ T-cell lymphopenia.
Interleukin-7 is a lymphoid cell growth factor which affects pre-B, pro-B and early T cells. It was first isolated by Namen in 1988. IL-7 has a number of known activities. It supports the growth of early B cells from long-term lymphoid bone marrow cultures. It is mitogenic for thymocytes and co-mitogenic with PHA and Con A. IL-7 also stimulates the proliferation of CD4/CD8 cells. The proliferative response of thymocytes to IL-7 is not affected by antibodies to the T cell growth factors such as IL-2, IL-4, and IL-6. Mature T cells respond to IL-7 and Con A, but not to IL-7 alone. Antibodies against IL-2 affect its activity, suggesting that it functions through IL-2 production. Human natural IL-7 is 25 kDa glycoprotein that has 6 cysteine residues which are important for biological activity. Human and mouse IL-7 have 60% amino acid sequence homology.
Lymphoid cell growth factor which affects pre-B, pro-B and early T cells; supports the growth of early B cells and is mitogenic for thymocytes and co-mitogenic with PHA and Con A; stimulates the proliferation of CD4/CD8 cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in 10 mM acetic acid containing 250 μg bovine serum albumin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianbao Gao et al.
International journal of molecular sciences, 16(5), 10267-10280 (2015-05-09)
Interleukin-7 (IL-7) is a non-hematopoietic cell-derived cytokine with a central role in the adaptive immune system. It promotes lymphocyte development in the thymus and maintains survival of naive and memory T cell homeostasis in the periphery. Moreover, it is important
Generalized verrucosis and HPV-3 susceptibility associated with CD4 T-cell lymphopenia caused by inherited human interleukin-7 deficiency.
Liran Horev et al.
Journal of the American Academy of Dermatology, 72(6), 1082-1084 (2015-05-20)
Fathollah Kalantar et al.
Iranian journal of immunology : IJI, 11(1), 1-12 (2014-03-19)
Type 2 diabetes (T2D) is a chronic metabolic disorder in which beta-cells are destroyed. The islet amyloid polypeptide (IAPP) produced by beta-cells has been reported to influence beta-cell destruction. To evaluate if IAPP can act as an autoantigen and therefore
Soichiro Saito et al.
Oncology reports, 16(5), 949-956 (2006-10-04)
Biliary tract cancers carry dismal prognoses. It is commonly understood that chromosomal aberrations in cancer cells have prognostic and therapeutic implications. However, in biliary tract cancers the genetic changes have not yet been sufficiently studied. The aim of this study
Craig A McElroy et al.
Structure (London, England : 1993), 17(1), 54-65 (2009-01-15)
IL-7 and IL-7Ralpha bind the gamma(c) receptor, forming a complex crucial to several signaling cascades leading to the development and homeostasis of T and B cells. We report that the IL-7Ralpha ectodomain uses glycosylation to modulate its binding constants to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service